83381 Sigma-Aldrich

Mercury(II) nitrate monohydrate

puriss. p.a., ACS reagent, ≥98.5% (T)

DOWNLOAD MSDS (PDF)

Synonym: Mercuric nitrate monohydrate

  • CAS Number 7783-34-8

  • Linear Formula Hg(NO3)2 · H2O

  • Molecular Weight 342.62

  •  EC Number 233-152-3

  •  MDL number MFCD00149736

  •  PubChem Substance ID 24887943

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, M-N, Puriss p.a. ACS, Puriss p.a. ACS More...
grade   ACS reagent
  puriss. p.a.
assay   ≥98.5% (T)
ign. residue (after reduction)   ≤0.01%
mp   79 °C(lit.)
anion traces   chloride (Cl-): ≤20 mg/kg
  sulfate (SO42-): ≤20 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  Hg+: ≤2000 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Price and Availability

Customers Also Viewed

Mercury(I) nitrate dihydrate

reagent grade, ≥97%

Mercury(II) nitrate monohydrate

≥99.99% trace metals basis

Mercury(II) nitrate monohydrate

ACS reagent, ≥98.0%

Mercury(II) nitrate solution

volumetric, 0.005 M Hg(NO3)2 (0.01N)

Mercury(II) nitrate solution

volumetric, 0.07 M Hg(NO3)2 (0.14N)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?