EMAIL THIS PAGE TO A FRIEND
83464 Sigma-Aldrich

RBS pF solution

concentrate

Purchase

Properties

Related Categories Cleaners, Detergents, Labware More...
impurities   ≤5% sodium hydroxide
density   1.07 g/mL at 20 °C

Description

Application

Special detergent for glass and instruments in chemical, bacteriological and radiotechnical laboratories; phosphate-free.

Other Notes

Sales restrictions may apply

Legal Information

RBS is a trademark of Chemical Products R. Borghgraef S.A.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?