• USA Home
  • 83544 - Mercury bromide paper

EMAIL THIS PAGE TO A FRIEND
83544 Sigma-Aldrich

Mercury bromide paper

for the detection of arsine

Synonym: Arsenic test paper

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Ion Tests, Test Papers/Sticks More...
quality   for the detection of arsine
size   20 mm × 70 mm

Description

Analysis Note

Limit of sensitivity: 0,5 μg As

Application

Determination of arsenic in grape must and wine, determination of arsine in air. In the latter case the test paper should first be moistened with acetic acid anhydride.

General description

This test paper is used for the determination of arsenic or arsine. It is baded on the fact that arsine forms a colored coordination compound with mercury compounds. The test paper is exposed to AsH3, liberated with zinc/hydrochloric acid, by holding the paper either over the test tube or over the end of a gas exhaust tube.

white → brown-black. The color reaction is faster, when the paper is first moistened before being used to test for AsH3 in air.

Packaging

Test paper box of 200 strips

Price and Availability

Safety & Documentation
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?