EMAIL THIS PAGE TO A FRIEND
83601 Sigma-Aldrich

Resorcinol

tested according to Ph.Eur.

Synonym: 1,3-Benzenediol, Resorcinolum

  • CAS Number 108-46-3

  • Linear Formula C6H4-1,3-(OH)2

  • Molecular Weight 110.11

  •  Beilstein Registry Number 906905

  •  EC Number 203-585-2

  •  MDL number MFCD00002269

  •  PubChem Substance ID 24888119

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z More...
vapor density   3.8 (vs air)
vapor pressure   1 mmHg ( 21.1 °C)
InChI Key   GHMLBKRAJCXXBS-UHFFFAOYSA-N
form   powder, flakes, or chunks
autoignition temp.   1126 °F
pharmacopeia   testing conforms to Pharmacopeia
bp   178 °C/16 mmHg(lit.)
mp   109-112 °C(lit.)
solubility   : soluble717 g/L at 25 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Blood, Central nervous system, Respiratory system
Personal Protective Equipment 
RIDADR 
UN 2876 6.1 / PGIII
WGK Germany 
2
RTECS 
VG9625000
Flash Point(F) 
260.6 °F
Flash Point(C) 
127 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?