• USA Home
  • 83639 - Resorufin β-D-glucopyranoside

EMAIL THIS PAGE TO A FRIEND
83639 Sigma-Aldrich

Resorufin β-D-glucopyranoside

for fluorescence, ≥95% (HPLC)

Synonym: 3-Phenoxazone 7-(β-D-glucopyranoside)

  • CAS Number 101490-85-1

  • Empirical Formula (Hill Notation) C18H17NO8

  • Molecular Weight 375.33

  •  Beilstein Registry Number 4335306

  •  MDL number MFCD00037662

  •  PubChem Substance ID 329769080

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Detection, Enzyme Substrates, Enzymes, Inhibitors, and Substrates,
grade   for fluorescence
InChI Key   QULZFZMEBOATFS-UYTYNIKBSA-N
assay   ≥95% (HPLC)
solubility   DMF: soluble
  DMSO: soluble
  H2O: soluble
fluorescence   λex 571 nm; λem 590 nm in 0.1 M phosphate pH 7.0 (β-glucosidase)
storage temp.   −20°C

Description

Application

suitable as fluorogenic substrate for β-glucosidase

Other Notes

Detection of β-glucosidase with fluorogenic probe

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability


Laboratory Gloves

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

R4758 Resorufin β-D-glucopyranoside, ≥90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?