• USA Home
  • 83725 - Rhodium(II) acetate dimer dihydrate

EMAIL THIS PAGE TO A FRIEND
83725 Sigma-Aldrich

Rhodium(II) acetate dimer dihydrate

purum, ~43% rhodium (Rh) basis

Synonym: Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Rhodium More...
InChI Key   SYBXSZMNKDOUCA-UHFFFAOYSA-J
grade   purum
assay   ~43% rhodium (Rh) basis

Description

Other Notes

Efficient catalyst for the cyclopropanation of olefins by diazo esters

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

209058 Rhodium(II) acetate dimer, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?