• USA Home
  • 83820 - D-(+)-Ribonic acid-γ-lactone

EMAIL THIS PAGE TO A FRIEND
83820 Sigma-Aldrich

D-(+)-Ribonic acid-γ-lactone

puriss., ≥99.0% (T)

Synonym: D-(+)-Ribono-1,4-lactone

  • CAS Number 5336-08-3

  • Empirical Formula (Hill Notation) C5H8O5

  • Molecular Weight 148.11

  •  Beilstein Registry Number 82057

  •  EC Number 226-256-5

  •  MDL number MFCD00063241

  •  PubChem Substance ID 329769088

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
InChI Key   CUOKHACJLGPRHD-BXXZVTAOSA-N
grade   puriss.
assay   ≥99.0% (T)
optical activity   [α]20/D +18.0±0.5°, c = 5% in H2O
mp   82-86 °C
solubility   H2O: 0.1 g/mL clear

Description

Other Notes

Chiral building block

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?