EMAIL THIS PAGE TO A FRIEND
83960 Sigma-Aldrich

Rubidium bromide

purum p.a., ≥99.0% (AT)

Synonym: Rubidium monobromide

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Inorganic Salts, Research Essentials, Rubidium,
InChI Key   JAAGVIUFBAHDMA-UHFFFAOYSA-M
grade   purum p.a.
assay   ≥99.0% (AT)
density   3.35 g/mL at 25 °C(lit.)
anion traces   sulfate (SO42-): ≤500 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤5000 mg/kg
  Na: ≤100 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?