• USA Home
  • 84186 - Lewatit® MonoPlus MP 64 free base

EMAIL THIS PAGE TO A FRIEND
84186 Sigma-Aldrich

Lewatit® MonoPlus MP 64 free base

Purchase

Properties

Related Categories Analytical/Chromatography, Anion Exchange Media, Lewatit Weak Anion Exchanger, Resins & LC Separation Media, Resins and Media More...
parameter   70 °C max. temp.
moisture   61-66%
loss   ~60% loss on drying, 110°C
matrix   styrene-divinylbenzene (macroporous)
matrix active group   tertiary/quaternary ammonium functional group
particle size   590 μm std dev ±50
operating pH   0 - 7
capacity   1.3 meq/mL by wetted bed volume
density   1.04 g/mL(lit.)
bulk density   660 g/L

Description

Analysis Note

Weakly basic anion exchange resin for demineralization and electroplating rinse water. The monodispersed particle size optimizes the kinetics necessary for best operating capacity.

Legal Information

Lewatit is a registered trademark of Lanxess Deutschland GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?