84380 Sigma-Aldrich

Nitric acid

puriss. p.a., ≥65% (T)

DOWNLOAD MSDS (PDF)
  • CAS Number 7697-37-2

  • Empirical Formula (Hill Notation) HNO3

  • Molecular Weight 63.01

  •  EC Number 231-714-2

  •  MDL number MFCD00011349

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, I-N, Puriss p.a., Puriss p.a. More...
vapor pressure   8 mmHg ( 20 °C)
grade   puriss. p.a.
assay   ≥65% (T)
ign. residue   ≤0.0005% (as SO4)
bp   120.5 °C(lit.)
density   1.37-1.41 g/mL at 20 °C(lit.)
anion traces   chloride (Cl-): ≤0.5 mg/kg
  phosphate (PO43-): ≤0.5 mg/kg
  sulfate (SO42-): ≤1 mg/kg
cation traces   Ag: ≤0.01 mg/kg
  Al: ≤0.05 mg/kg
  As: ≤0.01 mg/kg
  Ba: ≤0.01 mg/kg
  Bi: ≤0.1 mg/kg
  Ca: ≤0.5 mg/kg
  Cd: ≤0.01 mg/kg
  Co: ≤0.01 mg/kg
  Cr: ≤0.02 mg/kg
  Cu: ≤0.01 mg/kg
  Fe: ≤0.2 mg/kg
  Hg: ≤0.01 mg/kg
  K: ≤0.05 mg/kg
  Li: ≤0.01 mg/kg
  Mg: ≤0.1 mg/kg
  Mn: ≤0.01 mg/kg
  Mo: ≤0.02 mg/kg
  Na: ≤0.5 mg/kg
  Ni: ≤0.02 mg/kg
  Pb: ≤0.01 mg/kg
  Sr: ≤0.01 mg/kg
  Tl: ≤0.05 mg/kg
  Zn: ≤0.05 mg/kg

Price and Availability

Customers Also Viewed

Nitric acid

ACS reagent, 70%

Nitric acid

≥65%, TraceSELECT® Ultra, for ultratrace analysis

Nitric acid

puriss. p.a., 65% (Hg ≤0.0000005%), ≥65% (T)

Nitric acid

puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?