• USA Home
  • 84500 - (+)-Santolina alcohol

EMAIL THIS PAGE TO A FRIEND
84500 Sigma-Aldrich

(+)-Santolina alcohol

analytical standard

Synonym: (S)-2,5-Dimethyl-3-vinyl-4-hexen-2-ol

  • CAS Number 35671-15-9

  • Empirical Formula (Hill Notation) C10H18O

  • Molecular Weight 154.25

  •  Beilstein Registry Number 1902772

  •  MDL number MFCD00210090

  •  PubChem Substance ID 329769301

Purchase

Properties

Related Categories Additional Standards, Alcohols, Aliphatic, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   JWGLVEFPXSKNBN-VIFPVBQESA-N
assay   ≥98.5% (sum of enantiomers, GC)
form   neat
optical activity   [α]20/D +15.0±1°, neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.463
density   0.861 g/mL at 20 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?