EMAIL THIS PAGE TO A FRIEND
84530 Sigma-Aldrich

Sarcosine

≥99.0% (NT)

Synonym: N-Methylglycine

  • CAS Number 107-97-1

  • Linear Formula CH3NHCH2COOH

  • Molecular Weight 89.09

  •  Beilstein Registry Number 1699442

  •  EC Number 203-538-6

  •  MDL number MFCD00004279

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   FSYKKLYZXJSNPZ-UHFFFAOYSA-N
assay   ≥99.0% (NT)
ign. residue   ≤0.1% (as SO4)
loss   ≤0.1% loss on drying, 20 °C (HV)
mp   208-212 °C (dec.)(lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   As: ≤0.1 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

Solubilization of myelin proteins by detergents

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

S7672 Sarcosine, BioXtra

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?