EMAIL THIS PAGE TO A FRIEND
84650 Sigma-Aldrich

Sudan IV

for microscopy (Bot., Hist.)

Synonym: Biebrich scarlet R fat soluble, Fat Ponceau R or 4, Lipid Crimson, Oil Red IV, Scarlet Red Scharlach, Solvent Red 24, Sudan R or BB

  • CAS Number 85-83-6

  • Empirical Formula (Hill Notation) C24H20N4O

  • Molecular Weight 380.44

  •  Beilstein Registry Number 709018

  •  Colour Index Number 26105

  •  EC Number 201-635-8

  •  MDL number MFCD00003893

Purchase

Properties

Related Categories Biochemicals and Reagents, Luminescent Compounds/Detection, Stains and Dyes More...
grade   for microscopy (Bot., Hist.)
InChI Key   RCTGMCJBQGBLKT-PAMTUDGESA-N
mp   199 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QL5775000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?