EMAIL THIS PAGE TO A FRIEND
84683 Sigma-Aldrich

Sulfur

purum p.a., ≥99.5% (T)

Purchase

Properties

vapor density   8.9 (vs air)
vapor pressure   1 mmHg ( 183.8 °C)
  10 mmHg ( 246 °C)
grade   purum p.a.
assay   ≥99.5% (T)
autoignition temp.   450 °F
resistivity   2E23 μΩ-cm, 20°C
ign. residue   ≤0.2%
bp   444.7 °C(lit.)
mp   112.8 °C (rhombic)(lit.)
  117-120 °C(lit.)
  119.0 °C (monoclinic)(lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Na: ≤100 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Application

Sulfur has been employed as a reactant in the preparation of Cu-doped iron sulphides.

General description

Sulfur is a chalcogen (ore forming element). It is present in abundant quantities in marine and freshwater sediments. It exists in various forms such as pyrite, acid volatile sulfides, elemental sulfur, sulfates and organic sulfur. Quantitative estimation of reduced inorganic sulfur compounds in modern sediments and shales by a chromium reduction technique has been described.1 Elemental sulfur is employed for the synthesis of arsenic monosulfide and benzothiazole.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1350 4.1 / PGIII
WGK Germany 
1
Flash Point(F) 
404.6 °F
Flash Point(C) 
207 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?