EMAIL THIS PAGE TO A FRIEND
84712 Sigma-Aldrich

Carbon disulfide

purum, ≥99.0% (GC)

  • CAS Number 75-15-0

  • Empirical Formula (Hill Notation) CS2

  • Molecular Weight 76.14

  •  Beilstein Registry Number 1098293

  •  EC Number 200-843-6

  •  MDL number MFCD00011321

  •  PubChem Substance ID 329769310

Purchase

Properties

vapor density   2.67 (vs air)
InChI Key   QGJOPFRUJISHPQ-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
autoignition temp.   212 °F
expl. lim.   50 %
refractive index   n20/D 1.627(lit.)
  n20/D 1.627
bp   46 °C(lit.)
mp   −112-−111 °C(lit.)
solubility   water: soluble2.9 g/L at 20 °C
density   1.263 g/mL at 20 °C
  1.266 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1131 - class 3 - PG 1 - Carbon disulfide
WGK Germany 
2
RTECS 
FF6650000
Flash Point(F) 
-22 °F
Flash Point(C) 
-30 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?