EMAIL THIS PAGE TO A FRIEND
84714 Sigma-Aldrich

Carbon disulfide

puriss. p.a., ≥99.9% (GC)

  • CAS Number 75-15-0

  • Empirical Formula (Hill Notation) CS2

  • Molecular Weight 76.14

  •  Beilstein Registry Number 1098293

  •  EC Number 200-843-6

  •  MDL number MFCD00011321

  •  PubChem Substance ID 329769312

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, C-D, Puriss p.a., Puriss p.a. More...
vapor density   2.67 (vs air)
InChI Key   QGJOPFRUJISHPQ-UHFFFAOYSA-N
grade   puriss. p.a.
assay   ≥99.9% (GC)
autoignition temp.   212 °F
expl. lim.   50 %
impurities   ≤0.05% water
evapn. residue   ≤0.002%
refractive index   n20/D 1.627(lit.)
  n20/D 1.627
bp   46 °C(lit.)
mp   −112-−111 °C(lit.)
solubility   water: soluble2.9 g/L at 20 °C
density   1.263 g/mL at 20 °C
  1.266 g/mL at 25 °C(lit.)
anion traces   sulfate (SO42-): ≤2.5 mg/kg
  sulfite (SO32-): ≤10 mg/kg
cation traces   Al: ≤0.5 mg/kg
  Ba: ≤0.1 mg/kg
  Bi: ≤0.1 mg/kg
  Ca: ≤0.5 mg/kg
  Cd: ≤0.05 mg/kg
  Co: ≤0.02 mg/kg
  Cr: ≤0.02 mg/kg
  Cu: ≤0.02 mg/kg
  Fe: ≤0.1 mg/kg
  K: ≤0.5 mg/kg
  Li: ≤0.1 mg/kg
  Mg: ≤0.1 mg/kg
  Mn: ≤0.02 mg/kg
  Mo: ≤0.1 mg/kg
  Na: ≤0.5 mg/kg
  Ni: ≤0.1 mg/kg
  Pb: ≤0.1 mg/kg
  Sr: ≤0.1 mg/kg
  Zn: ≤0.1 mg/kg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1131 - class 3 - PG 1 - Carbon disulfide
WGK Germany 
2
RTECS 
FF6650000
Flash Point(F) 
-22 °F
Flash Point(C) 
-30 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?