• USA Home
  • 84940 - Semicarbazide hydrochloride

EMAIL THIS PAGE TO A FRIEND
84940 Sigma-Aldrich

Semicarbazide hydrochloride

purum p.a., ≥98.0% (AT)

Synonym: N-Aminourea hydrochloride, Hydrazine carboxamide hydrochloride

  • CAS Number 563-41-7

  • Linear Formula NH2CONHNH2 · HCl

  • Molecular Weight 111.53

  •  Beilstein Registry Number 3593642

  •  EC Number 209-247-0

  •  MDL number MFCD00013009

  •  PubChem Substance ID 329769327

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks, Ureas More...
InChI Key   XHQYBDSXTDXSHY-UHFFFAOYSA-N
grade   purum p.a.
assay   ≥98.0% (AT)
impurities   ≤0.05% hydrazine traces
ign. residue   ≤0.1%
mp   175-177 °C (dec.)(lit.)
solubility   H2O: soluble0.1 g/mL at 20 °C, clear, colorless
anion traces   sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Na: ≤100 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
VT3500000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?