EMAIL THIS PAGE TO A FRIEND
84960 Sigma-Aldrich

L-Serine

≥99.0% (NT)

Synonym: (S)-2-Amino-3-hydroxypropionic acid

  • CAS Number 56-45-1

  • Linear Formula HOCH2CH(NH2)CO2H

  • Molecular Weight 105.09

  •  Beilstein Registry Number 1721404

  •  EC Number 200-274-3

  •  MDL number MFCD00064224

Purchase

Properties

Related Categories Acacia, Allium cepa (Onion), Allium sativum (Garlic), Aloe Vera, Amino Acids & Derivatives,
InChI Key   MTCFGRXMJLQNBG-REOHCLBHSA-N
assay   ≥99.0% (NT)
optical activity   [α]20/D +13±1°, c = 5% in 5 M HCl
impurities   ≤0.5% foreign amino acids
ign. residue   ≤0.1% (as SO4)
loss   ≤0.2% loss on drying, 110 °C
mp   222 °C (dec.)(lit.)
solubility   H2O: very soluble359.7 g/L at 20 °C, clear (OECD Test Guideline 105)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  NH4+: ≤100 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Biochem/physiol Actions

Precursor of glycine by serine hydroxymethyltransferase.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

S4500 L-Serine, ReagentPlus®, ≥99% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?