• USA Home
  • 84999 - N-(7-Nitro-4-benzofurazanyl)-L-prolyl chloride

EMAIL THIS PAGE TO A FRIEND
84999 Sigma-Aldrich

N-(7-Nitro-4-benzofurazanyl)-L-prolyl chloride

for fluorescence

Synonym: (S)-(−)-1-(7-Nitro-4-benzofurazanyl)-2-pyrrolidinecarbonyl chloride, (S)-(−)-4-(2-Chloroformyl-pyrrolidino)-7-nitrobenzofurazan, (S)-(−)-NBD-Pro-COCl

  • CAS Number 159717-68-7

  • Empirical Formula (Hill Notation) C11H9ClN4O4

  • Molecular Weight 296.67

  •  MDL number MFCD03791269

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Benzofurazans, Carbonyl Chlorides, Derivatization Reagents,
grade   for fluorescence
fluorescence   λex 484 nm; λem 528 nm in acetonitrile
storage temp.   2-8°C

Description

Other Notes

Chiral fluorescent derivatizing reagent for the resolution of enantiomeric alcohols and amines by HPLC

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?