EMAIL THIS PAGE TO A FRIEND
85070 Sigma-Aldrich

Sesamol

purum, ≥98.0% (GC)

Synonym: 3,4-(Methylenedioxy)phenol, 5-Benzodioxolol

  • CAS Number 533-31-3

  • Empirical Formula (Hill Notation) C7H6O3

  • Molecular Weight 138.12

  •  Beilstein Registry Number 127405

  •  EC Number 208-561-5

  •  MDL number MFCD00005827

  •  PubChem Substance ID 329769355

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   LUSZGTFNYDARNI-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
mp   62-65 °C(lit.)
  62-65 °C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SM0890000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?