• USA Home
  • 85137 - Silver standard for AAS

EMAIL THIS PAGE TO A FRIEND
85137 Sigma-Aldrich

Silver standard for AAS

ready-to-use, traceable to BAM, in nitric acid

Synonym: Silver nitrate solution

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Matrix Selection,
InChI Key   SQGYOTSLMSWVJD-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   in nitric acid
  1.000 g/L Ag
application(s)   ICP/AAS: suitable
format   single component solution

Description

Preparation Note

prepared with AgNO3 and HNO3 (~0.5 M)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN3264 - class 8 - PG 3 - EHS - Corrosive liquid, acidic, inorganic, n.o.s., HI: all
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?