• USA Home
  • 85191 - Glutaraldehyde solution

EMAIL THIS PAGE TO A FRIEND
85191 Sigma-Aldrich

Glutaraldehyde solution

1.2 % (w/v) glutaraldehyde in H2O

Synonym: Glutaric dialdehyde solution, Pentane-1,5-dial, Silver stain kit component

  • CAS Number 111-30-8

  • Linear Formula OHC(CH2)3CHO

  • Molecular Weight 100.12

  •  Beilstein Registry Number 605390

  •  MDL number MFCD00007025

  •  PubChem Substance ID 329769361

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C1 to C6, Carbonyl Compounds, Cell Biology,
InChI Key   SXRSQZLOMIGNAQ-UHFFFAOYSA-N
concentration   1.2 % (w/v) glutaraldehyde in H2O
storage temp.   2-8°C

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?