EMAIL THIS PAGE TO A FRIEND
85208 Sigma-Aldrich

Silver ionophore II

Selectophore

Synonym: 6-Oxa-3,9-dithiabicyclo[9.3.1]pentadeca-1(15),11-13-triene, MAO

  • CAS Number 130184-19-9

  • Empirical Formula (Hill Notation) C12H16OS2

  • Molecular Weight 240.38

  •  Beilstein Registry Number 4182633

  •  MDL number MFCD00214192

  •  PubChem Substance ID 329769362

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   FFJVWROXKBHBGI-UHFFFAOYSA-N
mp   112-114 °C

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Silver ionophore used in solid PVC membrane electrode

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?