EMAIL THIS PAGE TO A FRIEND
85330 Sigma-Aldrich

Silica gel

technical grade, 1-3 mm particle size

Purchase

Properties

Related Categories Essential Chemicals, Other Silica Gels, Research Essentials, Silica Gels
grade   technical grade
InChI Key   VYPSYNLAJGMNEJ-UHFFFAOYSA-N
form   powder
particle size   1-3 mm
bp   2230 °C
mp   >1600 °C

Description

Packaging

1 kg in poly bottle

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?