• USA Home
  • 85351 - Silicon standard for AAS

EMAIL THIS PAGE TO A FRIEND
85351 Sigma-Aldrich

Silicon standard for AAS

ready-to-use, in HNO3: HF

Synonym: Silicate solution

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Matrix Selection,
InChI Key   XUIMIQQOPSSXEZ-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   in HNO3: HF
  1.000 g/L Si
application(s)   ICP/AAS: suitable
format   single component solution

Description

Preparation Note

prepared with Si and HNO3 (~1 M) and 2% HF

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2922 6.1(8) / PGII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

16259 Silicon Standard for AAS, TraceCERT®, 1000 mg/L Si in NaOH

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?