EMAIL THIS PAGE TO A FRIEND
85382 Sigma-Aldrich

GC Stationary Phase

phase OV-25, bottle of 10 g

Purchase

Properties

Related Categories Analytical/Chromatography, GC Stationary Phases, Gas Chromatography (GC), Packed GC Columns and Components
grade   for GC
description   a methylphenylsilicon (-75% phenyl)
packaging   bottle of 10 g
operating limits   ≤305 °C
application(s)   gas chromatography (GC): suitable
matrix active group   OV-25 phase

Description

General description

Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?