• USA Home
  • 85434 - Silylating mixture I according to Sweeley

EMAIL THIS PAGE TO A FRIEND
85434 Sigma-Aldrich

Silylating mixture I according to Sweeley

for GC derivatization, ampule

Synonym: Chlorotrimethylsilane – hexamethyldisilazane mixture, HMDS + TMCS, Hexamethyldisilazane/TMCS 2/1 (v/v) in pyridine

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents GC, Reagents for Silylation,
grade   for GC derivatization
description   ready to use solution in pyridine (10 vol parts)
InChI Key   UCJNZOHHMSKSRP-UHFFFAOYSA-N
quality   ampule
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1992 6.1(3) / PGII
WGK Germany 
2
Flash Point(F) 
62.6 °F
Flash Point(C) 
17 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

Silylating Reagents for GC-MS Compatible Derivatives of NSAIDs

Human and veterinary pharmaceuticals are a group of “emerging” contaminants,1, 2 some of which are produced in increasingly large volumes every year. The amounts produced are reaching quantities simi...
Luis-Alberto Martin
AnalytiX Volume 10 Article 2
Keywords: Anti-inflammatory agents, Derivatizations, Environmental, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Metabolites, Pesticides, Pharmaceutical, Silylations, Veterinary

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?