EMAIL THIS PAGE TO A FRIEND
85438 Sigma-Aldrich

SIM Medium

for microbiology

Synonym: Sulfide Indole Motility Medium

Purchase

Properties

Related Categories All Food Control Media (alphabetical), All Liquid Media/Broths, All Solid Media/Agars, Biochemical Identification Media by Test, Biochemical Identification Media, A to Z,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 3.0 g/L
  ammonium ferric citrate, 0.2 g/L
  casein peptone, 20.0 g/L
  meat peptone, 6.6 g/L
  sodium thiosulfate, 0.2 g/L
final pH   7.3±0.2 (25 °C)
suitability   Providencia
  nonselective and differential for Citrobacter
  nonselective and differential for Enterobacteriaceae
  nonselective and differential for Enterococcus
  nonselective and differential for Escherichia coli and Coliforms
  nonselective and differential for Klebsiella
  nonselective and differential for Proteus
  nonselective and differential for Salmonella
  nonselective and differential for Shigella

Description

Application

Sulfide indole motility (SIM) medium is a test agar for Enterobacteriaceae, for the detection of motility and the formation of sulfide and indole.

General description

Sulfide indole motility (SIM) medium is a semi-solid medium containing cysteine and iron ions. Darkening of the medium (a black precipitate) indicates the presence of bacteria producing hydrogen sulfide.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Differentiation and Identification Media for Salmonella

Salmonella contamination is the second leading cause of food-borne illness worldwide. Controlling outbreaks of Salmonella is an important task for food regulators, restaurants and the food industry i...
Jvo Siegrist
Microbiology Focus Edition 1.3
Keywords: Biochemistry, Fermentation, Food & Beverage, Indicators, Microbiology

Differentiation of Escherichia coli from coliforms

In August 2008, the discovery of E. coli-contaminated beef in the United States prompted a nationwide recall of beef. The source turned out to be one supplier that had a history of contamination of i...
Jvo Siegrist
AnalytiX Volume 8 Article 5
Keywords: Decarboxylations, Fermentation, Genetic, Indicators, Microbiology, Reductions

EZ-Product Family for Bioburden Testing

The EZ family of products has been designed to optimize and streamline the bioburden workflow. The family includes:
Keywords: Filtration, Separation

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?