EMAIL THIS PAGE TO A FRIEND
85530 Sigma-Aldrich

D-Sorbitol

crystallized, ≥99.0% (HPLC)

Synonym: D-Glucitol

  • CAS Number 50-70-4

  • Empirical Formula (Hill Notation) C6H14O6

  • Molecular Weight 182.17

  •  Beilstein Registry Number 1721899

  •  EC Number 200-061-5

  •  MDL number MFCD00004708

Purchase

Properties

Related Categories Aloe Vera, Carbohydrate Chemistry, Carbohydrate Derivatives, Carbohydrates, Cell Biology,
vapor density   <1 (vs air)
vapor pressure   <0.1 mmHg ( 25 °C)
InChI Key   FBPFZTCFMRRESA-JGWLITMVSA-N
assay   ≥99.0% (HPLC)
form   powder or crystals
optical activity   [α]20/D −1.5±0.3°, c = 10% in H2O
quality   crystallized
impurities   ≤1.0% water
ign. residue   ≤0.1% (as SO4)
mp   ~95 °C
  98-100 °C(lit.)
solubility   H2O: soluble0.1 g/mL, clear, colorless
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   As: ≤0.1 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Biochem/physiol Actions

D-Sorbitol is a sugar alcohol that is commonly used as a sugar substitute. It occurs naturally and is also produced synthetically from glucose. The food industry uses D-sorbitol as an additive in the form of a sweetener, humectant, emulsifier, thickener, or dietary supplement. D-Sorbitol has also been found in cosmetics, paper, and pharmaceuticals. Naturally, D-sorbitol occurs widely in plants via photosynthesis, ranging from algae to higher order fruits of the family Rosaceae.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
LZ4290000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

85529 D-Sorbitol, BioUltra, ≥99.5% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?