EMAIL THIS PAGE TO A FRIEND
85544 Sigma-Aldrich

Span® 20

Synonym: Sorbitan monolaurate, Span® 20

  • CAS Number 1338-39-2

  • Empirical Formula (Hill Notation) C18H34O6

  • Molecular Weight 346.46

  •  Beilstein Registry Number 8355657

  •  EC Number 215-663-3

  •  MDL number MFCD00005365

  •  PubChem Substance ID 329769383

Purchase

Properties

Related Categories Biochemicals and Reagents, Cleaners, Detergents, Detergents N to Z, Labware,
description   non-ionic
InChI Key   LWZFANDGMFTDAV-WYDSMHRWSA-N
refractive index   n20/D 1.474(lit.)
HLB value   8.6±1.0
density   1.032 g/mL at 25 °C(lit.)

Description

Legal Information

Span is a registered trademark of Croda International PLC

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WG2920000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?