• USA Home
  • 85570 - Spermidine phosphate salt hexahydrate

EMAIL THIS PAGE TO A FRIEND
85570 Sigma-Aldrich

Spermidine phosphate salt hexahydrate

≥98.0% (TLC)

Synonym: Dispermidine triphosphate

  • CAS Number 23273-82-7

  • Linear Formula [NH2(CH2)3NH(CH2)4NH2]2 · 3H3PO4 · 6H2O

  • Molecular Weight 346.28

  •  MDL number MFCD04966492

  •  PubChem Substance ID 329769387

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Enzyme Inhibitors, Enzyme Inhibitors by Type,
InChI Key   UYNKOICVVIGVMD-UHFFFAOYSA-N
assay   ≥98.0% (TLC)
impurities   ≤0.2% spermine
storage temp.   2-8°C

Description

Biochem/physiol Actions

Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA, may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity.

Caution

Solutions should be sterile-filtered, not autoclaved, if sterile solution is necessary. Spermidine deaminates with time; solutions should be stored frozen. Prepare new solutions frequently.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?