EMAIL THIS PAGE TO A FRIEND
85680 Sigma-Aldrich

Stearic acid

puriss., ≥98.5% (GC)

Synonym: 1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

  • CAS Number 57-11-4

  • Linear Formula CH3(CH2)16COOH

  • Molecular Weight 284.48

  •  Beilstein Registry Number 608585

  •  EC Number 200-313-4

  •  MDL number MFCD00002752

  •  PubChem Substance ID 329769401

Purchase

Price and Availability


Laboratory Gloves

The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
WI2800000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S4751 Stearic acid, Grade I, ≥98.5% (capillary GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?