• USA Home
  • 85793 - Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) complex

EMAIL THIS PAGE TO A FRIEND
85793 Sigma-Aldrich

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) complex

for fluorescence, ≥95.0% (HPCE)

  • CAS Number 123148-15-2

  • Empirical Formula (Hill Notation) C72H48F12N6P2Ru

  • Molecular Weight 1388.19

  •  MDL number MFCD03791108

  •  PubChem Substance ID 24888456

Purchase

Description

Other Notes

Physical properties

Same product — same quality — now Sigma-Aldrich brand

Packaging

A protocol for embedding polystyrene beads is included with the material.

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Tris(4,7-diphenyl-1,10-phenanthroline)-ruthenium hexafluorophosphate (Ru(dpp)3(PF6)2) has been widely used as a probe for luminescent detection and quantitation of oxygen. The fluorescence of the dye (abs. λmax 455 nm, luminescence λ max 613 nm) is strongly reduced by molecular oxygen due to dynamic quenching, making Ru(dpp)3(PF6)2 a viable oxygen probe based on either measurement of intensity or decay time. At a partial pressure of 200 hPa the decay time (and intensity) of Ru(dpp)3(PF6)2 decreases by a factor of 2 due to dynamic quenching. Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium hexafluorophosphate is recommended for conjugation to polystyrene membranes or beads.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?