• USA Home
  • 85874 - Streptococcus Selective Agar

EMAIL THIS PAGE TO A FRIEND
85874 Sigma-Aldrich

Streptococcus Selective Agar

for microbiology

Purchase

Properties

Related Categories All Solid Media/Agars, Clinical Media, Enterococcus, Media, Media by Application,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 13 g/L
  casein peptone, 14.4 g/L
  crystal violet, 0.0002 g/L
  L-cystine, 0.2 g/L
  D(+)-glucose, 5 g/L
  sodium azide, 0.2 g/L
  sodium chloride, 4 g/L
  sodium citrate, 1 g/L
  sodium sulfite, 0.2 g/L
  soy peptone, 5 g/L
final pH   7.4±0.2 (25 °C)
suitability   selective for Enterococcus
  selective for Streptococcus

Description

Application

For the selective enrichment and isolation of streptococci from all types of material, in particular material grossly contaminated with accompanying flora, according to the studies of Pike.

Price and Availability

Safety & Documentation

Safety Information

Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?