EMAIL THIS PAGE TO A FRIEND
85875 Sigma-Aldrich

trans-Stilbene oxide

purum, ≥98.0% (HPLC)

Synonym: trans-1,2-Diphenyloxirane

  • CAS Number 1439-07-2

  • Empirical Formula (Hill Notation) C14H12O

  • Molecular Weight 196.24

  •  Beilstein Registry Number 82740

  •  EC Number 215-877-7

  •  MDL number MFCD00064311

  •  PubChem Substance ID 329769420

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Epoxides, Organic Building Blocks, Oxygen Compounds More...
InChI Key   ARCJQKUWGAZPFX-ZIAGYGMSSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   65-67 °C(lit.)
  66-69 °C

Description

Other Notes

This product has been replaced by S4921-ALDRICH | trans-Stilbene oxide 98%

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DT4391500

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of trans-Stilbene Oxide Enantiomers on Astec® Cellulose DMP

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of trans-Stilbene Oxide Enantiomers on Astec® Cellulose DMP
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S4921 trans-Stilbene oxide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?