• USA Home
  • 85884 - Streptomycin sulfate salt

EMAIL THIS PAGE TO A FRIEND
85884 Sigma-Aldrich

Streptomycin sulfate salt

tested according to Ph.Eur.

Synonym: Streptomycini sulfas

  • CAS Number 3810-74-0

  • Linear Formula C21H39N7O12 · 1.5 H2SO4

  • Molecular Weight 728.69

  •  Beilstein Registry Number 3894995

  •  EC Number 223-286-0

  •  MDL number MFCD00037023

  •  PubChem Substance ID 329769422

Purchase

Properties

Related Categories Aminoglycosides, Antibacterial, Antibiotics, Antibiotics A to Z, Antibiotics N-S,
InChI Key   QTENRWWVYAAPBI-YZTFXSNBSA-N
form   solid
pharmacopeia   testing conforms to Pharmacopeia
potency   ≥720 I.U. per mg (Calc. on dry substance)
shelf life   ~4 yr
mol wt   mol wt 728.69 g/mol
pH   4.5-7.0
anion traces   sulfate (SO42-): 18.0-21.5% (Calculated on dry substance)
Mode of action   protein synthesis | interferes
antibiotic activity spectrum   Gram-negative bacteria
  Gram-positive bacteria

Description

Application

Streptomycin is used to study mechanisms of streptomycin resistance and is often used together with penicillin and other agents to inhibit bacterial contamination in cell culture applications. It is recommended for use in molecular biology applications at 25-50μg/mL, in cell culture applications at 100 mg/L and in embryo culture at 50 mg/L.

Biochem/physiol Actions

Mode of Action: Streptomycin acts by inhibiting prokaryote protein synthesis by binding to the S12 protein of the 30S ribosomal subunit, thus preventing the transition from imitation complex to chain-elongating ribosome. This causes miscoding or inhibits initiation.

Mode of Resistance: A mutation in rpsL, a gene for S12 ribosomal protein, prevents binding of streptomycin to the ribosome. An aminoglycoside phosphotransferase also inactivates streptomycin.

Antimicrobial spectrum: Streptomycin acts against gram-negative and gram-positive bacteria.

Caution

Solutions can be stored at 2-8°C for up to a month or at -20°C for more extended periods. At 37°C, solutions are stable for three days.

Preparation Note

Stock solutions should be prepared directly in the vial with sterile water at concentrations between 20-25 mg/mL.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
WK4990000

Documents

Certificate of Analysis

Protocols & Articles

Articles

Inhibition of Protein Synthesis by Antibiotics

Protein synthesis is a complex, multi-step process involving many enzymes as well as conformational alignment. However, the majority of antibiotics that block bacterial protein synthesis interfere wi...
BioFiles 2006, 1.4, 17.
Keywords: Antibiotics, Environmental, Peptide synthesis

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?