EMAIL THIS PAGE TO A FRIEND
86018 Sigma-Aldrich

Sulfacetamide

≥98.0% (NT)

Synonym: N-(4-Aminobenzenesulfonyl)acetamide, N-(4-Aminophenylsulfonyl)acetamide, N1-Acetylsulfanilamide

  • CAS Number 144-80-9

  • Empirical Formula (Hill Notation) C8H10N2O3S

  • Molecular Weight 214.24

  •  Beilstein Registry Number 981718

  •  EC Number 205-640-6

  •  MDL number MFCD00066501

  •  PubChem Substance ID 329769433

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics N-S, Biochemicals and Reagents, Building Blocks,
InChI Key   SKIVFJLNDNKQPD-UHFFFAOYSA-N
assay   ≥98.0% (NT)
ign. residue   ≤0.4%
mp   182-184 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AC8450000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?