• USA Home
  • 86129 - Sodium 3-sulfobenzoate

EMAIL THIS PAGE TO A FRIEND
86129 Sigma-Aldrich

Sodium 3-sulfobenzoate

purum, ≥98.0% (T)

Synonym: 3-Carboxybenzenesulfonic acid sodium salt, 3-Sulfobenzoic acid sodium salt

  • CAS Number 17625-03-5

  • Linear Formula NaO3SC6H4CO2H

  • Molecular Weight 224.17

  •  Beilstein Registry Number 5675222

  •  EC Number 241-602-5

  •  MDL number MFCD00066378

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acid Salts, Sulfur Compounds More...
InChI Key   KQHKITXZJDOIOD-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (T)
impurities   ≤1% water
mp   ≥300 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?