• USA Home
  • 86182 - N-(3-Sulfopropyl)-3,3′,5,5′-tetramethylbenzidine sodium salt

EMAIL THIS PAGE TO A FRIEND
86182 Sigma-Aldrich

N-(3-Sulfopropyl)-3,3′,5,5′-tetramethylbenzidine sodium salt

≥98.0% (HPLC)

Synonym: 4-Amino-4′-sulfopropylamino-3,3′,5,5′-tetramethylbiphenyl sodium salt, TMBZ.PS

Purchase

Properties

InChI Key   JOBKFCDDANZFOP-UHFFFAOYSA-M
assay   ≥98.0% (HPLC)

Description

Application

Chromogenic reagent for the detection of hydrogen peroxide e.g. in the enzymatic analysis of body fluids.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?