EMAIL THIS PAGE TO A FRIEND
86257 Sigma-Aldrich

Talc

tested according to Ph.Eur.

Synonym: Hydrous magnesium silicate, Talcum

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z More...
grade   Ph Eur
InChI Key   FPAFDBFIGPHWGO-UHFFFAOYSA-N
pharmacopeia   testing conforms to Pharmacopeia

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
WW2710000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?