• USA Home
  • 86311 - Bis(pentafluorophenyl)phenylphosphine

EMAIL THIS PAGE TO A FRIEND
86311 Sigma-Aldrich

Bis(pentafluorophenyl)phenylphosphine

analytical standard, for mass spectrometry

Synonym: 2,3,4,5,6,2′,3′,4′,5′,6′-Decafluorotriphenylphosphine

  • CAS Number 5074-71-5

  • Linear Formula C6H5P(C6F5)2

  • Molecular Weight 442.19

  •  Beilstein Registry Number 6253897

  •  EC Number 225-780-1

  •  MDL number MFCD00000291

  •  PubChem Substance ID 24888583

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, MS Markers, MS Standards,
grade   analytical standard, for mass spectrometry
InChI Key   OYNXPGGNQMSMTR-UHFFFAOYSA-N
form   neat
mp   55-60 °C(lit.)
  55-60 °C
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?