EMAIL THIS PAGE TO A FRIEND
86410 Sigma-Aldrich

Terephthalaldehyde

purum, ≥98.0% (HPLC)

Synonym: Benzene-1,4-dicarboxaldehyde, Terephthaldicarboxaldehyde

  • CAS Number 623-27-8

  • Linear Formula C6H4(CHO)2

  • Molecular Weight 134.13

  •  Beilstein Registry Number 385863

  •  EC Number 210-784-8

  •  MDL number MFCD00006949

  •  PubChem Substance ID 24888646

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C8, Carbonyl Compounds, Chemical Synthesis,
InChI Key   KUCOHFSKRZZVRO-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
impurities   ≤1% water
bp   245-248 °C(lit.)
mp   114-116 °C(lit.)
  114-116 °C

Description

Other Notes

This product has been replaced by T2207-ALDRICH | Terephthalaldehyde ReagentPlus, 99%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WZ0430000
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

T2207 Terephthalaldehyde, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?