• USA Home
  • 86611 - Tetraethylammonium chloride

EMAIL THIS PAGE TO A FRIEND
86611 Sigma-Aldrich

Tetraethylammonium chloride

≥98.0% (AT)

Synonym: TEA chloride

  • CAS Number 56-34-8

  • Linear Formula (C2H5)4N(Cl)

  • Molecular Weight 165.70

  •  Beilstein Registry Number 3563247

  •  EC Number 200-267-5

  •  MDL number MFCD00011828

  •  PubChem Substance ID 329769473

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   YMBCJWGVCUEGHA-UHFFFAOYSA-M
assay   ≥98.0% (AT)
impurities   ≤2% water

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BS6125000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?