• USA Home
  • 86612 - Tetraethylammonium bromide

EMAIL THIS PAGE TO A FRIEND
86612 Sigma-Aldrich

Tetraethylammonium bromide

puriss., ≥99.0% (AT)

Synonym: TEA bromide

  • CAS Number 71-91-0

  • Linear Formula (C2H5)4N(Br)

  • Molecular Weight 210.16

  •  Beilstein Registry Number 3563430

  •  EC Number 200-769-4

  •  MDL number MFCD00011825

  •  PubChem Substance ID 329769474

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   HWCKGOZZJDHMNC-UHFFFAOYSA-M
grade   puriss.
assay   ≥99.0% (AT)
mp   285 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
BS5950000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?