• USA Home
  • 86613 - Tetraethylammonium cyanide

EMAIL THIS PAGE TO A FRIEND
86613 Sigma-Aldrich

Tetraethylammonium cyanide

purum, ≥95% (AT)

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   PCZOZSATUTWXIC-UHFFFAOYSA-N
grade   purum
assay   ≥95% (AT)
mp   250 °C (dec.)(lit.)

Description

Other Notes

Reagent for the synthesis of nitriles

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN2811 - class 6.1 - PG 2 - MP - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?