• USA Home
  • 86620 - Tetraethylammonium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
86620 Sigma-Aldrich

Tetraethylammonium tetrafluoroborate

purum, ≥98.0% (T)

  • CAS Number 429-06-1

  • Linear Formula (C2H5)4N(BF4)

  • Molecular Weight 217.06

  •  Beilstein Registry Number 3917638

  •  EC Number 207-055-1

  •  MDL number MFCD00011827

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   XJRAKUDXACGCHA-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   ≥300 °C(lit.)

Description

Other Notes

This product has been replaced by 242144-ALDRICH | Tetraethylammonium tetrafluoroborate 99%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

242144 Tetraethylammonium tetrafluoroborate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?