• USA Home
  • 86630 - Tetraethylammonium hydroxide solution

EMAIL THIS PAGE TO A FRIEND
86630 Sigma-Aldrich

Tetraethylammonium hydroxide solution

technical, ~20% in H2O

Synonym: TEA hydroxide

  • CAS Number 77-98-5

  • Linear Formula (C2H5)4N(OH)

  • Molecular Weight 147.26

  •  Beilstein Registry Number 3912961

  •  MDL number MFCD00009024

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Synthetic Reagents More...
InChI Key   LRGJRHZIDJQFCL-UHFFFAOYSA-M
grade   technical
concentration   ~20% in H2O
impurities   ≤3% halides (as chloride)
anion traces   sulfate (SO42-): ≤1000 mg/kg

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3267 8 / PGII
WGK Germany 
3
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

177806 Tetraethylammonium hydroxide solution, 20 wt. % in H2O

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?