• USA Home
  • 86640 - Tetraethylammonium iodide

EMAIL THIS PAGE TO A FRIEND
86640 Sigma-Aldrich

Tetraethylammonium iodide

puriss., ≥98.5% (CHN)

  • CAS Number 68-05-3

  • Linear Formula (C2H5)4N(I)

  • Molecular Weight 257.16

  •  Beilstein Registry Number 3562649

  •  EC Number 200-676-9

  •  MDL number MFCD00011829

  •  PubChem Substance ID 329769484

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   UQFSVBXCNGCBBW-UHFFFAOYSA-M
grade   puriss.
assay   ≥98.5% (CHN)
impurities   ≤1% water
mp   >300 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BS7365000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

235938 Tetraethylammonium iodide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?