• USA Home
  • 86691 - Tetraethylene glycol di(p-toluenesulfonate)

EMAIL THIS PAGE TO A FRIEND
86691 Sigma-Aldrich

Tetraethylene glycol di(p-toluenesulfonate)

purum, ≥95.0% (HPLC), colorless to yellow

Synonym: Tetraethylene glycol ditosylate

  • CAS Number 37860-51-8

  • Linear Formula (CH3C6H4SO3CH2CH2OCH2CH2)2O

  • Molecular Weight 502.60

  •  Beilstein Registry Number 1610992

  •  MDL number MFCD00075239

  •  PubChem Substance ID 24888715

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Tosylates More...
InChI Key   SLAONPBUWDUSSO-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (HPLC)
color   colorless to yellow
refractive index   n20/D 1.53(lit.)
density   1.242 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?