EMAIL THIS PAGE TO A FRIEND
86770 Sigma-Aldrich

Tetrabromomethane

purum, ≥97.0% (GC)

Synonym: Carbon tetrabromide

  • CAS Number 558-13-4

  • Empirical Formula (Hill Notation) CBr4

  • Molecular Weight 331.63

  •  Beilstein Registry Number 1732799

  •  EC Number 209-189-6

  •  MDL number MFCD00000117

  •  PubChem Substance ID 329769488

Purchase

Properties

Related Categories Bromination, C-X Bond Formation (Halogen), Chemical Synthesis, Synthetic Reagents
vapor pressure   40 mmHg ( 96 °C)
InChI Key   HJUGFYREWKUQJT-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
bp   190 °C(lit.)
mp   88-90 °C(lit.)
  92-93 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2516 6.1 / PGIII
WGK Germany 
3
RTECS 
FG4725000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C11081 Tetrabromomethane, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?